missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Hopen™ Single Use Hygiene Gown

Product Code. 16524081 Shop All Medicom Products
Change view
Click to view available options
Closure Type:
Press Studs
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type Size Unit Size
16524081 Press Studs Medium Pack of 50
16534081 Press Studs Large Pack of 50
16544081 Press Studs X-Large Pack of 50
16554081 Press Studs 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16524081 Supplier Medicom™ Supplier No. 40.550/02
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medicom™ Hopen™ Single Use Hygiene Gown without pockets, is microporous and waterproof

  • Reduces contamination of the operator, but also sensitive products and processes in a protected atmosphere
  • Protects against radioactive particles and infectious agents
  • Microporous, Anti-Static Hygiene Gown is comfortable and light
  • Polypropylene gown lined with laminated polyethylene for a waterproof, hydrophobic seal
  • Available in mulitple sizes, and closure types

Specifications

Product Type General Purpose Gown
Size Medium
Gender Unisex
Material Polypropylene
Color White
Closure Type Press Studs
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Radioactive Particles, Infectious Agents
No. of Pockets None
Finish Microporous, Waterproof
Seam Type Sewn
Certifications/Compliance ISO 9001
Certifications/Compliance (Europe) EN 1149-5
For Use With (Application) Category 1 Personal Protective Equipment
Packaging Type Individual, Bag
Autoclavable Not Autoclavable
Construction Made in China
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.