missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Hopen™ Pocketless Hygiene Gown

Product Code. 16524051 Shop All Medicom Products
Change view
Click to view available options
Color:
Red
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size Unit Size
16524051 White Medium Pack of 50
17790109 Red X-Large Pack of 50
17700119 Red 2X-Large Pack of 50
17710119 Red 3X-Large Pack of 50
16534051 White Large Pack of 50
16544051 White X-Large Pack of 50
16554051 White 2X-Large Pack of 50
16564051 White 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 16524051 Supplier Medicom™ Supplier No. 40.301/02
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medicom™ Hopen™ Pocketless Hygiene Gown available in three distinct colors

  • The Pocketless Hygiene gown protects the working environment from particles coming from the body of the operator
  • Allows the user to be protected from the risks of soiling related to the activity being carried out
  • Pleasant and light, polypropylene fabric is extremely breathable
  • Collared model without flap and elasticated at the wrists

Specifications

Size Medium
Construction Made in China
For Use With (Application) Category 1 Personal Protective Equipment
Gender Unisex
No. of Pockets None
Packaging Type Individual, Bag
Product Type Hygiene Gown
Material Polypropylene
Color White
Closure Type Press Studs
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Seam Type Sewn
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.