Learn More
Nabertherm Muffle Furnaces
Nabertherm furnaces are perfect all-rounder for research and laboratory applications. They are manufactured from expertly finished, high quality materials and are easy to operate. The insulation out of non-classified fibre materials permits energysaving operation and fast heating times due to low heat storage and thermalconductivity.
Brand: Nabertherm L-032K1RN1
Code : NEW
Additional Details : Weight : 20.00000kg
Description
- Heating from two sides by ceramic heating plates with integrated heating element which is safeguarded against fumes and splashing, and easy to replace
- Insulation made of non-classified fibre material
- Textured stainless steel sheet housing
- Equipped with either a flap door (L) or lift door (LT)
- Dual shell housing for low external temperatures and high stability
- Adjustable air inlet integrated into the door, exhaust air outlet in the rear wall of furnace
- Supplied with B410 controller with four programs and five freely programmable ramps, or C450 with 10 freely adjustable programs and 20 segments
- Both controllers have integrated USB interface and offer plain text display in different languages
- Alternative furnace sizes and temperature ranges are available on request