missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ iBind™ Flex Western Device
Description
The iBind™ Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.
The Invitrogen™ iBind™ Flex Western System offers:
The Invitrogen™ iBind™ Flex Western System offers:
- Flexible options—optimize antibody use, change formats and reduce hands-on time
- Cost savings—use up to 80% less primary antibody
- Reproducibility—automated processing provides improved blot-to-blot consistency
Specifications
Specifications
| Capacity | 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots |
| Detection Method | Fluorescence, Chemiluminescence, Chromogenic |
| For Use With (Application) | Western Blot |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | iBind Flex |
| Running Time | ∼2.5 hr. |
| Throughput | 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots |
| Type | Western Device |
| Includes | Roller |
| Quantity | 1 device |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
