Learn More
Zeiss™ Primovert Inverted Microscope with Phototube
Product Code. 15168229
Change view
Click to view available options
Objective Types:
4X and 10X
4X, 10X, 20X and 40X
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Objective Types unitSize
15168229 4X, 10X, 20X and 40X Each
10636717 4X and 10X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15168229 Supplier Zeiss™ Supplier No. 4912060011000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Compact inverted routine microscope that combines efficiency and optical performance. This inverted microscope is configured for transmitted light brightfield and phase contrast. This system is ideal for cell culture, routine work and quick & easy imaging.
  • The Primovert is perfectly suited to your cell culture laboratory.
  • It enables fast, efficient investigations of both unstained cells in phase contrast and GFP-labeled cells in fluorescence contrast.
  • It fits straight into your laminar flow cabinet to work directly in a sterile environment.
  • The Primovert is easy to use and with little set up the system is ready to use out of the box.
  • Automatic light source switch-off (in addition to manual on/off-function): Primo Vert switches off the light automatically after 15 min - saving energy and protecting the stand
  • Modular illumination: halogen or long lasting LED
  • High quality material: the stand is made of metal and all operating elements are coated with a special touch-friendly rubber compound
  • Universal phase slider (optional) for all objectives: for the ease of use there is one phase ring (Ph1) available for 10x, 20x and 40x - this means not having to change the position of the phase slider when changing objectives. In short: time-saving and economical
  • Increased working distance thanks to easy removal of the condenser, e.g., for roller-bottles
  • Objective indicators for fast identification of magnification
  • Integrated carrying handle
  • Award-winning design The new Primo Vert inverted microscope is focused on the essentials: excellent optics, high-durability quality materials, ease of use and an appealing design.
  • The Primo Vert addresses a wide range of applications: from routine laboratories for live cell inspection to cutting edge research laboratories which require - in addition to research microscopes - compact and reliable routine microscopes for quick and efficient viewing of living cells.
  • Typical applications include cancer and HIV research, human, animal and plant genetics, and cell biology in general.
Specifications
Includes Plan Achromat LD 20 x/0.3 Ph1 and LD 40 x/0.5 Ph1
Objective Types 4X, 10X, 20X and 40X
For Use With (Application) Cell Culture, Routine Work and Quick and Easy Imaging