Learn More
Thermo Scientific™ Pierce™ Fast Blocking Buffer
Shop All Thermo Scientific ProductsDescription
Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with Thermo Scientific™ Pierce Fast Blocking Buffer, a ready-to-use blocker solution.
Additional Information and Online Ordering
Highlights:
Fast – use both buffers to shorten the typical Western blot development by over 2 hours
Simple – optimized protocol makes Western blot analysis easier than ever
Low background – provides results comparable to classic Western blotting buffers
Specifications
Specifications
| Chemical Name or Material | Blocking Buffer |
| Concentration | 1X |
| For Use With (Application) | Western Blot |
| Physical Form | Liquid |
| Quantity | 100 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.