missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Spectra™ Multicolor High Range Protein Ladder

Product Code. 11872124 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2 x 250 μL
8 x 250 μL
Unit Size:
Pack of 2
Pack of 4
This item is not returnable. View return policy

Product Code. 11872124

Brand: Thermo Scientific™ 26625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A mixture of eight blue-, green- and orange-stained proteins (40 to 300kDa) for use as size standards for high MW proteins in gel electrophoresis and Western blotting.

Thermo Scientific™ Spectra Multicolor High Range Protein Ladder is a mixture of eight (8) blue-, green and orange-stained proteins (40 to 300kDa) for use as size standards for high MW proteins in gel electrophoresis and Western blotting.

This prestained protein MW marker is designed for monitoring the progress of SDS-polyacrylamide gel electrophoresis, for assessing transfer efficiency onto PVDF, nylon and nitrocellulose membranes, and for estimating the approximate size of separated proteins that have been made visible with gel stains or Western blot detection reagents. Three different chromophores (blue, orange, green) are bound to the different component proteins, producing a brightly colored ladder with an easy-to-remember pattern. The Spectra Multicolor High Range Protein Ladder is ready to use: no heating, further dilution or addition of a reducing agent is required.

Highlights:

  • Size range – eight proteins spanning 40 to 300kDa

  • Multicolor – three different colors for unambiguous band-size assignment

  • Ready-to-use – supplied in a loading buffer for direct loading on gels; no need to boil

  • Sharp bands – color-coded bands of similar intensity for easy visualization

  • Quality tested – each lot evaluated by SDS-PAGE and Western blotting

  • Membrane-compatible – colored bands transfer to membranes for Western blotting

Includes:

Dye-stained proteins in 62.5mM Tris-H3PO4 (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 10mM DTT, 1mM NaN3 and 33% glycerol.

Recommended for:

Monitoring of protein migration during SDS-PAGE; Verifying Western transfer efficiency; Approximate sizing of proteins on SDS-polyacrylamide gels and Western blots; Locating a protein of interest for excision from an unstained preparative gel

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contents: two vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

Detection Method Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm)
Number of Markers 8
Product Type Protein Ladder
Ready to Load Yes
Recommended Applications Tris-acetate gels
Size Range 40 to 300 kDa
Stain Type 3 colors: Blue, Orange, Green
For Use With (Application) Tris-acetate gels
Gel Compatibility Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 300, 250, 180, 130, 100, 70, 50, 40 kDa
Product Line Spectra
Quantity 2 x 250 μL
Shipping Condition Approved for shipment on Wet or Dry Ice
System Type Western Blotting, SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.