missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Penetration Electrodes

Featuring a longer life than a conventional electrode, this gel electrolyte electrode is ideal for meat, cheese, fruit, bread dough.

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.08000kg

Product Code. 11850980

  • £126.14 / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

  • Gel electrolyte electrode for meat, cheese, fruit, paste, bread
  • Longer life than a conventional electrode
Specifications

Specifications

Penetrating gel electrolyte electrode
Penetration Electrode
Gel
0°C to 60°C
8 mm
120 mm
2 to 13
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers