missing translation for 'onlineSavingsMsg'
Learn More

Shamrock™ Tyvek™ Tag

Product Code. 11812651
Click to view available options
Dimensions (LxW):
25 x 51 mm
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11812651

Brand: Shamrock™ STV3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides extra strength, adhesion and permanent marking for your laboratory needs. Shamrock™ Tyvek™ Tags won't discolor, crack or tear and adhere to almost any surface.

  • Tear-proof material, long-lasting adhesive
  • Does not discolor over time
  • Permanent marking with ballpoint pen, paper pencil and felt marker
  • Labels pre-cut on roll

Specifications

Product Type Label Tyvek Tag
Quantity 1000/Pk.
Dimensions (LxW) 25 x 51 mm
Length (Metric) 25 mm
Width (Metric) 51 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.