missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
Plunger, 47.5g; weights, 50 and 100g
- Precision machined and ruggedly constructed to ensure long service life in both laboratory and field applications
- Full penetration range of 0 to 62mm with 1/10mm subdivisions (0 to 620 penetration scale)
- Accommodates cones and needles to perform all of the ASTM tests on lubricating greases, asphalts, petroleum waxes and petrolatums
- Compact design allows for easy transport
- Head assembly adjusts for accurate placement of the tip of the needle or cone on the surface of the sample
- Sturdy cast-iron base provides excellent support and has a built-in spirit level and level screws to ensure proper alignment of the penetrometer during testing
- L × W × H: 15 × 15 × 46cm (6 × 6 × 18 in.)
Specifications
Specifications
| Includes | Plunger, Weights |
| Weight (English) | 12 lb. |
| Weight (Metric) | 5.4 kg |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?
