Learn More
Fisherbrand™ Epoxy Corps Amber pH Electrode
Offered in a variety of styles including different reference solutions and reference chambers; these electrodes feature a refillable outer reference reservoir that can be filled with any electrolyte compatible with the sample, including those fills recommended for non-aqueous measurements.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.18000kg
Description
- Double junction reference is recommended for samples containing organic compounds, proteins, heavy metals or compounds that interact with silver (e.g. sulfide, bromide, iodide)
- All electrodes are available with a detachable lead or with a cable/connector attached
- Designed for all existing Brinkmann, Metrohm and Schott titration systems
- The electrode body is constructed with a glass ground joint that will fit their titration vessels
- Ionics and Tytronics titrators can be fitted with this same electrode without the ground joint
- Either design can be ordered depending on the titration unit
Specifications
Epoxy Corps Amber pH Electrode | |
Electrode |