missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Clear Autoclave Bags
Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.60000kg
Description
- For high temperature sterilisation (134°C) to a maximum of 140°C
- 40μm thickness
- High temperature polypropylene bags for the decontamination and inactivation of particularly resistant biological waste
- For safe dissposal of biohazard waste
- Specifically designed for contaminated waste disposal in autoclaves or incinerators
- Printed with a biohazard warning