missing translation for 'onlineSavingsMsg'
Learn More

Guanosine, MP Biomedicals™

Product Code. 11374359 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 11374359

Brand: MP Biomedicals 0210190780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 118-00-3
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00010182
InChI Key NYHBQMYGNKIUIF-UUOKFMHZSA-N
Synonym guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
PubChem CID 6802
ChEBI CHEBI:16750
SMILES NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Color White
Quantity 100 g
IUPAC Name 2-amino-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Formula Weight 283.2
Physical Form Powder
Chemical Name or Material Guanosine
compliance-icons
Product Identifier
  • Guanosine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Guanosine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.