missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaclear™ Centrifugal Filters

Product Code. 10783307 Shop All Sartorius Products
Click to view available options
Pore Size:
0.8 μm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10783307

Brand: Sartorius VK01P042

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

500 - 100μL For fast and reliable clarification / filtration.

Vivaclear centrifugal filters are disposable microfiltration units for the fast and reliable clarification/filtration of biological samples in the range from 100 to 500μL. Vivaclear units feature a high-flux PES membrane of 0.8μm pore size and can be used in fixed angle rotors accepting 2.2 ml centrifuge tubes. Standard applications are sample clarification before loading onto Vivapure protein purification spin columns, particle removal, plasma or serum filtration, and removal of cells or cell debris from cell lysates or cell supernatants.

  • High-flux, 0.8 mm PES membrane
  • Used in fixed angle rotors
  • Low hold-up volume
  • Fast and reproducible performance
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
Pore Size 0.8 μm
Sample Volume 0.1 to 0.5 mL
Sterility Non-sterile
Quantity 100/Pk.
Volume (Metric) 0.5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.