Learn More
Applied Biosystems™ Dynabeads™ MX MPC™-6 Semi-automated Mixer Head
Dynabeads™ MX MPC™-6 Semi-automated Mixer Head
Brand: Applied Biosystems™ 15914D
Code : NEW
Additional Details : Weight : 0.00450kg
Description
Attachment for Dynabeads™ MX Mixer Base (Cat. No. 159-02). Designed to hold one MPC™-6 (Cat. No. 120-02D) sample rack; capacity up to 6 tubes.Specifications
Dynabeads™ MX MPC™-6 Semi-automated Mixer Head, 1 Unit, Sample Mixer Accessories Type, Designed to Hold one MPC™-6 Sample Rack; Capacity up to 6 Microcentrifuge Tubes, Use With Dynabeads™ Sample Mixers, Ideal for Food and Beverage | |
Food and beverage, industrial and applied science | |
Dynabeads™ sample mixers | |
1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.