missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts
GREENER_CHOICEBIOPHARMA

Product Code. 10615345 Shop All Corning Products
Click to view available options
Pore Size:
0.4 μm
3 μm
5 μm
8 μm
No. of Wells:
6
12
24
Diameter (Metric) Outer:
6.5 mm
12 mm
24.5 mm
Culture Area:
0.33 cm2
1.12 cm2
4.7 cm2
Unit Size:
Pack of 24
Pack of 48
This item is not returnable. View return policy

Product Code. 10615345

Brand: Corning™ 3402

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
  • Pore sizes ranging from 0.4 μm to 8 μm diameters
  • Treated for optimal cell attachment
  • Supplied in multiple well plates
  • Membrane must be stained for cell visibility
  • Sterile
TRUSTED_SUSTAINABILITY

Specifications

Description 12 Inserts
Culture Area 1.12 cm2
Material Polycarbonate
Volume (Metric) Well 1.5 mL
Pore Size 3 μm
Type Pre-loaded Microporous Insert in Tissue Culture Plate
For Use With (Application) Epithelial and endothelial cell culture; transport, migration/invasion, co-culture, and microbial pathogenesis studies
Diameter (Metric) Outer 12 mm
No. per Pack 4 plates; 12 inserts/plate
Surface Coating TC-Treated
Color Clear
Surface Treatment TC-Treated
Sterility Sterile
No. of Wells 12
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.