Learn More
Applied Biosystems™ Dynabeads™ MX MPC™-S Mixing Wheel
Dynabeads™ MX MPC™-S Mixing Wheel
Brand: Applied Biosystems™ 15911
Code : NEW
Additional Details : Weight : 0.40000kg
Description
Attachment for Dynabeads™ MX Mixer Base (Cat. No. 159-02). Designed to hold two MPC™-S sample racks (Cat. No. A13346); capacity up to 12 microcentrifuge tubes.Specifications
Mixing Wheel | |
Dynabeads Sample Mixer |
For Research Use Only. Not for use in diagnostic procedures.