Learn More
Thermo Scientific™ Abgene™ 96 Well 1.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Optimize suspension cell culture with these U-bottomed 1.2 mL storage plates.
Brand: Thermo Scientific™ AB-0787
249.20 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : 04
Additional Details : Weight : 6.70000kg
Description
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
Thermo Scientific 1.2 mL Storage Plates come with either round or square shapes, and they are made of polypropylene. These plates are ideal for suspension cell culture and are compatible with automated systems. Different sealing options are available for the 1.2 mL Storage Plates, and you can seal each well independently to prevent cross-contamination.
Highlights
• Round well shape designed for optimal sample recovery or square shape to maximize sample volume within ANSI footprint design
• Each well has an independent sealing rim to prevent cross-contamination
• U-bottomed wells ideally suited for sample resuspension
• Polypropylene (AB-1068, polystyrene)
• Resistant to DMSO (100%); Ethanol (100%); Isopropanol (100%)
• Suitable for Autoclaving (15 minutes at 121°C) or Gamma Irradiation
•Sealing options: Adhesive Seals, Heat Seals, Storage Plate Caps and Cap Strips, and Storage Plate Sealing Mats
• Cleanroom manufactured
• ANSI-format for compatibility with automated systems
Applications
• Sample storage
• Sample resuspension
• Cell culture
• Assay setup: diluting and aliquoting
Compatible Sealing Film:Adhesive Heat
> Hazardous:No
> Max Centrifugal Force:2000 g (AB-0564 and AB-1127)
> Max Vol (Cap Seal):1.1 mL (AB-0564 only)
> Max Vol (Film Seal):1.2 mL
> Max Vol (Mat Seal):0.85 mL (AB-0564, AB-1068), 1 mL (AB-1127)
> Storage Condition:Ambient
Sample storage, sample resuspension, cell culture and assay setup: diluting and aliquoting.
Certifications
Free from human DNA, RNases, and DNases.
Specifications
Custom Barcoding Available | |
96-well | |
8 x 12 | |
Bulk | |
Abgene | |
96-well plate | |
Polypropylene | |
96 | |
Cleanroom | |
Round | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
50 | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
96-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Natural | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
1200 μL | |
1000 μL | |
Non-treated | |
Yes | |
2000 x g | |
50 | |
Yes | |
ANSI/ SLAS Standard |