Learn More
Applied Biosystems™ BeadRetriever™ Tube Rack
BeadRetriever™ Tube Rack
Brand: Applied Biosystems™ 15952
Code : NEW
Additional Details : Weight : 0.31000kg
Description
Tube-sample rack, designed of use with the BeadRetriever™ System (Cat. No 159-50). The Tube Rack contains fifteen separate sample tube slots. Tubes for sample processing on the BeadRetriever™ System available separately (Cat. No. 159-51).Specifications
Tube Rack | |
Food and beverage, industrial and applied science | |
1 Each |
Tube Rack, 1 Unit, Use With the BeadRetriever™ System, Store at Room Temperature, Contains Fifteen Separate Sample Tube Slots, For Sample Processing, For Research Use Only, Not for Use in Diagnostic Procedures | |
BeadRetriever™ system |
For Research Use Only. Not for use in diagnostic procedures.