missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ Q Chromatography Media

Product Code. 10324120 Shop All Cytiva Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10324120

Brand: Cytiva 17531602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for capture and intermediate purification of proteins from large feed volumes. Cytiva CaptoTM Q Chromatography Media is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers.

  • Rigid, high flow agarose matrix modified with dextran surface extenders and strong quaternary ammonium (Q) anion exchanger
  • Combination of high volume throughput and dynamic binding capacity at high flow for raise in productivity
  • Process time reduced due to combination of high capacity with high flow velocity
  • Withstands effective CIP procedure and is chemically stable
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Stability 1 M Acetic Acid, 1 M NaOH, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
For Use With (Application) For Efficient Capture and Intermediate Protein PurifiCatIon When High Volume Throughput is Essential
Storage Requirements 4°C to 30°C
Product Type Capto Q Media
Flow Rate 700 cm/Hoμr
Particle Size 90 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Ionic Capacity 0.16 to 0.22 mM Cl-/mL
Ion Exchange Type Strong AnIon Exchanger
Max. Pressure 0.3 MPa
Matrix Highly Cross-Linked Agarose with Dextran surface extender
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type Quaternary Amine
Show More Show Less
compliance-icons
Product Identifier
  • Capto Q, 100 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.