missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis
116.42 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
55.20 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
55.20 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Use promo code "23271" to get your promotional price.
55.20 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
55.20 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Description
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis
Specifications
pH | 12 |
Boiling Point | 120°C |
Melting Point | 8°C |
Quantity | 5 L |
Packaging | HDPE plastic bottle |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'