missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, Certified AR for Analysis, Stabilised with BHT, meets analytical specification of Ph.Eur, Fisher Chemical™
43.65 GBP valid until 2024-05-31
Use promo code "21688" to get your promotional price.
43.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
43.72 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "21688" to get your promotional price.
43.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
43.72 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Aluminum Can
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Description
C4H10O, CAS Number-60-29-7, ether, pronarcol, 3-oxapentane, anesthetic ether, aether, anaesthetic ether, diethyl oxide, diethyl ether, ethyl ether, ethyl oxide, 2.5L, 34.6 deg.C, CHEBI:35702, Colorless, 74.12g/mol, RTZKZFJDLAIYFH-UHFFFAOYSA-N, ethoxyethane, -116 deg.C, 11646, 74.123
Specifications
Melting Point | -116°C |
Color | Colorless |
Boiling Point | 34.6°C |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 587mbar at 20°C |
Viscosity | 0.2448cp at 20°C |
Formula Weight | 74.12g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'