missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Closures for Dropper Bottles: Bulk Pack
258.92 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Use promo code "23291" to get your promotional price.
Click to view available options
Neck Finish:
15-415
Color:
Blue
Green
Natural
Red
White
Yellow
Description
Order Thermo Scientific™ Nalgene™ Closures for Dropper Bottles to complete the Nalgene Dropper Bottle System. Closures fit dropper bottles Cat. Nos. 312750 and 31275, and are available in multiple colors to help identify reagents.
Features:
- Polypropylene
- Available in various colors
Ordering Information:
Dropper Bottle Closures are bulk packed 2000 per case. Order Nalgene controlled-drop dispensing tip (Cat. Nos. 312758, 312759) and dropper bottles (Cat. Nos. 312750, 312751) separately.
Specifications
Closure Material | Polypropylene |
For Use With (Equipment) | Dropper Bottles 312750 and 312751. |
Product Type | Closure |
Color | Red |
Material | Polypropylene |
No. per Case | 2000 |
No. per Pack | 2000 |
Neck Finish | 15-415 |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. |
Closure Included | Yes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'