Learn More
Applied Biosystems™ Dynabeads™ anti-Listeria
Dynabeads anti-Listeria
Brand: Applied Biosystems™ 71006
Code : 50
Additional Details : Weight : 0.50000kg
Description
For the selective enrichment of Listeria directly from pre-enrichment samples using ImmunoMagnetic Separation. Method can be performed manually or can be automated by using a BeadRetriever™.Benefits and features
Saves processing and confirmation time
This Product Contains: Dynabeads™ coated with anti-Listeria antibodies
Specifications
Store in refrigerator (2–8°C). | |
Dynabeads™ MPC™, KingFisher mL Food Protection Purification System | |
Dynabeads™ |
Antibody Coated Bead | |
Small Volume Microbial Samples | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.