missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid Glacial, Certified AR for analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
34.92 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
35.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
35.73 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "21664" to get your promotional price.
35.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
35.73 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
Description
C2H4O2, CAS Number-64-19-7, acetasol, acide acetique, essigsaeure, vinegar acid, ethanoic acid, acetic acid, glacial, methanecarboxylic acid, glacial, ethylic acid, 2.5L, 117 deg.C, CHEBI:15366, Colorless, 60.05g/mol, QTBSBXVTEAMEQO-UHFFFAOYSA-N, 16 deg.C, 36152, 60.052, <-0.1ppm
Specifications
pH | 2.5 |
Melting Point | 16°C |
Boiling Point | 117°C |
Color | Colorless |
Formula Weight | 60.05g/mol |
Quantity | 1 L |
Packaging | HDPE plastic bottle |
Physical Form | Liquid |
Vapor Density | 2.1 |
Vapor Pressure | 1.52kPa at 20°C |
Safety and Handling
Product Identifier
- Acetic acid
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'