missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ PVDF BNC Connector Combination pH Electrode

Brand:  Hanna Instruments™ FC200B

Code : 04

Additional Details : Weight : 0.25000kg

Product Code. 10142801

  • £254.00 / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

BNC
Used on any pH meter that has the female BNC probe input
0 to 12
BNC Connector Combination pH Electrode
PVDF Body
0°C to 50°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers