missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylene dichloride, 98%

Product Code. 11471728
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471728 25 g 25g
11481728 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471728 Supplier Thermo Scientific Alfa Aesar Supplier No. A13976.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-25-6 | C8H8Cl2 | 175.052 g/mol

p-Xylylene dichloride is a monomer used in preparation of poly(p-phenylene vinylene(PPV) nanotubes and nanorods by chemical vapor deposition (CVD) polymerization method. It was used in a study to investigate the effect of CVD monomer selection and reaction conditions on the resulting PPV polymer layer composition and luminescence properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylylene dichloride is a monomer used in preparation of poly(p-phenylene vinylene(PPV) nanotubes and nanorods by chemical vapor deposition (CVD) polymerization method. It was used in a study to investigate the effect of CVD monomer selection and reaction conditions on the resulting PPV polymer layer composition and luminescence properties.

Solubility
Insoluble in water

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-25-6
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000920
InChI Key ZZHIDJWUJRKHGX-UHFFFAOYSA-N
Synonym 1,4-bis chloromethyl benzene, alpha,alpha'-dichloro-p-xylene, p-xylylene dichloride, p-xylylene chloride, benzene, 1,4-bis chloromethyl, p-bis chloromethyl benzene, benzene, p-bis chloromethyl, a,a'-dichloro-p-xylene, xylylene dichloride, ccris 1772
PubChem CID 12171
IUPAC Name 1,4-bis(chloromethyl)benzene
SMILES C1=CC(=CC=C1CCl)CCl

Specifications

Melting Point 98°C to 102°C
Boiling Point 254°C
Quantity 25 g
UN Number UN2928
Beilstein 774706
Solubility Information Insoluble in water
Formula Weight 175.06
Percent Purity 98%
Chemical Name or Material p-Xylylene dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.