missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylene dibromide, 97%

Product Code. 11471668
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11471668

Brand: Thermo Scientific Alfa Aesar A13939.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-24-5 | C8H8Br2 | 263.96 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in water (react), and toluene.

Notes
Incompatible with alcohols, bases, amines, oxidizing agents. Avoid moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-24-5
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000182
InChI Key RBZMSGOBSOCYHR-UHFFFAOYSA-N
Synonym 1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
PubChem CID 69335
IUPAC Name 1,4-bis(bromomethyl)benzene
SMILES C1=CC(=CC=C1CBr)CBr

Specifications

Melting Point 144°C to 148°C
Density 2.012
Boiling Point 245°C
Quantity 50 g
UN Number UN3261
Beilstein 1100019
Solubility Information Soluble in water (react),and toluene.
Formula Weight 263.96
Percent Purity 97%
Chemical Name or Material p-Xylylene dibromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.