missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Xylene, ReagentPlus™, 99%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
2.5 L
20 L
500mL
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | 12°C |
| Color | Colorless |
| Density | 0.861g/mL at 20°C |
| Boiling Point | 137°C to 139°C |
| Quantity | 2.5 L |
| Explosion Limit | 0.07 |
| Flash Point | 25°C (77°F) |
| Ignition Point | 984°F |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction