missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylene-d4 (ring-d4), CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | p-Xylene ring-d4 |
| CAS | 16034-43-8 |
| Molecular Formula | C8 2H4 H6 |
| Quantity | 0.5 g |
| Synonym | p-Xylene D4 (Ring D4), p-Xylene-2,3,5,6-d4 (6 CI,8 CI), 3,6-Dimethylbenzene-1,2,4,5-d4, 1,2,4,5-Tetradeuterio-3,6-dimethylbenzene, Benzene-1,2,4,5-d4, 3,6-dimethyl- |
| InChI Formula | InChI=1 S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6 H,1-2H3/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(C)c([2 H])c([2 H])c1C |
| IUPAC Name | 1,2,4,5-tetradeuterio-3,6-dimethylbenzene |
| Molecular Weight (g/mol) | 110.19 |
| Formula Weight | 110.1034 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction