missing translation for 'onlineSavingsMsg'
Learn More

p-Tyramine, TRC

Artikelnummer. 30133578
Ansicht ändern
Click to view available options
Quantity:
5 g
25 g
50 g
Packungsgröße:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30133578 25 g 25g
30133569 50 g 50g
17927524 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133578 Supplier Toronto Research Chemicals Supplier No. TRCT89850025G
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-67-2
Molecular Formula C8 H11 N O
Molecular Weight (g/mol) 137.18
Synonym Tyramine, Ritodrine Hydrochloride Imp. A (BP), Phenol, 4-(2-aminoethyl)-, Phenol, p-(2-aminoethyl)- (8CI), 4-(2-Aminoethyl)phenol, 2-(4-Hydroxyphenyl)ethanamine, 2-(4-Hydroxyphenyl)ethylamine, 2-(4'-Hydroxyphenyl)ethylamine, 2-(p-Hydroxyphenyl)ethylamine, 4-Hydroxy-β-phenylethylamine, 4-Hydroxyphenethylamine, 4-Hydroxyphenylethylamine, Benzeneethanamine, 4-hydroxy-, NSC 249188, Systogene, Tocosine, Tyramin, Tyrosamine, Uteramine, p-(2-Aminoethyl)phenol, p-Hydroxy-β-phenylethylamine, p-Hydroxyphenethylamine, p-Hydroxyphenylethylamine, p-Tyramine, p-β-Aminoethylphenol, α-(4-Hydroxyphenyl)-β-aminoethane, β-(4-Hydroxyphenyl)ethylamine
IUPAC Name 4-(2-aminoethyl)phenol
SMILES NCCc1ccc(O)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H11NO/c9-6-5-7-1-3-8(10)4-2-7/h1-4,10H,5-6,9H2
Formula Weight 137.0841
Percent Purity >95%
Chemical Name or Material Tyramine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.