missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylurea, 98+%, Thermo Scientific™

Product Code. 11499063
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499063 100 g 100g
11489063 25 g 25g
11409073 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499063 Supplier Thermo Scientific Alfa Aesar Supplier No. A11150.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-51-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00025433
InChI Key DMSHKWHLXNDUST-UHFFFAOYSA-N
Synonym p-tolylurea, p-tolylcarbamide, 4-tolylurea, 4-methylphenyl urea, urea, p-tolyl, 1-p-tolyl urea, 1-4-methylphenyl urea, p-tolycarbamide, urea, 4-methylphenyl, n-4-methylphenyl urea
PubChem CID 12148
IUPAC Name (4-methylphenyl)urea
SMILES CC1=CC=C(C=C1)NC(=O)N

Specifications

Melting Point 177°C to 184°C
Quantity 100 g
Beilstein 2208088
Formula Weight 150.18
Percent Purity ≥98%
Chemical Name or Material p-Tolylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.