missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylhydrazine hydrochloride, 98%

Product Code. 11492317
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492317 100 g 100g
11472317 5 g 5g
11482317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492317 Supplier Thermo Scientific Alfa Aesar Supplier No. A14220.22
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 637-60-5 | C7H11ClN2 | 158.629 g/mol

p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.

Solubility
Soluble in water, methanol and dimethyl sulfoxide.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-60-5
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD00012940
InChI Key HMHWNJGOHUYVMD-UHFFFAOYSA-N
Synonym p-tolylhydrazine hydrochloride, 4-methylphenylhydrazine hydrochloride, 4-methylphenyl hydrazine hydrochloride, 4-tolylhydrazine hydrochloride, hydrazine, 4-methylphenyl-, monohydrochloride, p-tolylhydrazinium 1+ chloride, hydrazine, p-tolyl-, hydrochloride, 4-methylphenylhydrazine hcl, 1-4-methylphenyl hydrazine hydrochloride, p-tolyl-hydrazine hydrochloride
PubChem CID 12504
IUPAC Name (4-methylphenyl)hydrazine;hydrochloride
SMILES CC1=CC=C(C=C1)NN.Cl

Specifications

Melting Point ∼240°C (decomposition)
Quantity 100 g
UN Number UN2811
Beilstein 3563996
Solubility Information Soluble in water,methanol and dimethyl sulfoxide.
Formula Weight 158.63
Percent Purity 98%
Chemical Name or Material p-Tolylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.