missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetylene, 98%

Product Code. 11406576
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11406576

Brand: Thermo Scientific Alfa Aesar B24812.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 766-97-2 | C9H8 | 116.163 g/mol

4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, heavy metal powders.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-97-2
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00008571
InChI Key KSZVOXHGCKKOLL-UHFFFAOYSA-N
Synonym 4-ethynyltoluene, p-tolylacetylene, 4-methylphenylacetylene, p-ethynyltoluene, p-methylphenylacetylene, benzene, 1-ethynyl-4-methyl, toluene, p-ethynyl, benzene, 1-ethynyl-4-methyl-9ci, 4'-methylphenyl acetylene, 1-ethynyl-4-methyl-benzene
PubChem CID 13018
IUPAC Name 1-ethynyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)C#C

Specifications

Melting Point ∼21°C
Density 0.916
Boiling Point 168°C to 170°C
Flash Point 49°C (120°F)
Odor Pungent
Refractive Index 1.547
Quantity 25 g
UN Number UN3295
Beilstein 969653
Solubility Information Insoluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material p-Tolylacetylene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.