missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetyl chloride, ≥97%, Thermo Scientific™

Product Code. 15427857
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427857 250 mg 250mg
15417857 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427857 Supplier Thermo Scientific Alfa Aesar Supplier No. H37997.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35675-44-6
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00018813
InChI Key QDZAWVLWIMOXJT-UHFFFAOYSA-N
Synonym p-tolylacetyl chloride, 2-4-methylphenyl acetyl chloride, p-tolyl-acetyl chloride, 4-methyl-phenyl-acetyl chloride, 4-methylphenylacetylchloride, p-methylphenylacetyl chloride, 2-4-tolyl acetyl chloride, 4-methylphenylacetyl chloride, 4-methylphenyl acetyl chloride, 4-methylphenylacetic acid chloride
PubChem CID 2760634
IUPAC Name 2-(4-methylphenyl)acetyl chloride
SMILES CC1=CC=C(CC(Cl)=O)C=C1

Specifications

Color Pink
Boiling Point 115°C (7.5 mmHg)
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 168.62
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material p-Tolylacetyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.