missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetonitrile, 98+%

Product Code. 11434803
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11434803

Brand: Thermo Scientific Alfa Aesar A12169.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2947-61-7 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2947-61-7
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00001922
InChI Key RNHKXHKUKJXLAU-UHFFFAOYSA-N
Synonym 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile
PubChem CID 76280
IUPAC Name 2-(4-methylphenyl)acetonitrile
SMILES CC1=CC=C(CC#N)C=C1

Specifications

Melting Point 18°C
Density 0.989
Boiling Point 242°C to 243°C
Flash Point 106°C (222°F)
Refractive Index 1.519
Quantity 5 g
UN Number UN3276
Beilstein 1099645
Formula Weight 131.18
Percent Purity ≥98%
Chemical Name or Material p-Tolylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.