missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetonitrile, 98+%

Product Code. 11454803
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454803 100 g 100g
11434803 5 g 5g
11444803 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454803 Supplier Thermo Scientific Alfa Aesar Supplier No. A12169.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2947-61-7 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2947-61-7
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00001922
InChI Key RNHKXHKUKJXLAU-UHFFFAOYSA-N
Synonym 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile
PubChem CID 76280
IUPAC Name 2-(4-methylphenyl)acetonitrile
SMILES CC1=CC=C(CC#N)C=C1

Specifications

Melting Point 18°C
Density 0.989
Boiling Point 242°C to 243°C
Flash Point 106°C (222°F)
Refractive Index 1.519
Quantity 100 g
UN Number UN3276
Beilstein 1099645
Formula Weight 131.18
Percent Purity ≥98%
Chemical Name or Material p-Tolylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.