missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetic acid, 99%

Product Code. 10351294 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351294 100 g Glass bottle 100g
10296283 5 g Glass bottle 5g
10745981 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10351294 Supplier Thermo Scientific Acros Supplier No. 139121000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-47-9 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-47-9
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00004353
InChI Key GXXXUZIRGXYDFP-UHFFFAOYSA-N
Synonym p-tolylacetic acid, 4-methylphenylacetic acid, 2-p-tolyl acetic acid, 2-4-methylphenyl acetic acid, p-methylphenylacetic acid, 4-tolylacetic acid, 4-methylphenyl acetic acid, benzeneacetic acid, 4-methyl, acetic acid, p-tolyl, 2-p-tolylacetic acid
PubChem CID 248474
IUPAC Name 2-(4-methylphenyl)acetic acid
SMILES CC1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 90°C to 93°C
Color White
Boiling Point 265°C to 267°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4CH2CO2H
Quantity 100 g
Beilstein 09, 530
Formula Weight 150.18
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material p-Tolylacetic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • p-Tolylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.