missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl isocyanate, 99%

Product Code. 10193790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193790 25 g Glass bottle 25g
10559063 5 g Glass bottle 5g
10205930 100 g Glass bottle 100g
11925681 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10193790 Supplier Thermo Scientific Acros Supplier No. 162520250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-58-2 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-58-2
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
InChI Key MGYGFNQQGAQEON-UHFFFAOYSA-N
Synonym p-tolyl isocyanate, 4-tolyl isocyanate, 4-methylphenyl isocyanate, p-isocyanatotoluene, 4-isocyanatotoluene, p-toluene isocyanate, p-methylphenyl isocyanate, benzene, 1-isocyanato-4-methyl, p-tolylisocyanate, 1-isocyanato-4-methyl-benzene
PubChem CID 69325
ChEBI CHEBI:53732
IUPAC Name 1-isocyanato-4-methylbenzene
SMILES CC1=CC=C(C=C1)N=C=O

Specifications

Color Brown
Density 1.0560g/mL
Boiling Point 70.0°C to 72.0°C (10.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4NCO
Refractive Index 1.5301 to 1.5321
Quantity 25 g
Beilstein 12,955
Specific Gravity 1.056
Formula Weight 133.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material p-Tolyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Tolyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.