missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl chlorothionoformate, 97%

Product Code. 11495507
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495507 50 g 50g
11485507 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495507 Supplier Thermo Scientific Alfa Aesar Supplier No. B23501.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 937-63-3 | C8H7ClOS | 186.65 g/mol

p-Tolyl chlorothionoformate is used in dehydration reactions by cyclic elimination of thiocarbonate esters. It acts as a catalyst in the synthesis of alfa-L-2'-deoxythreofuranosyl nucleoside analogs and alkenes from hindered alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolyl chlorothionoformate is used in dehydration reactions by cyclic elimination of thiocarbonate esters. It acts as a catalyst in the synthesis of alfa-L-2′-deoxythreofuranosyl nucleoside analogs and alkenes from hindered alcohols.

Solubility
Miscible with common organic solvents.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937-63-3
Molecular Formula C8H7ClOS
Molecular Weight (g/mol) 186.65
MDL Number MFCD00014466
InChI Key UNCAXIZUVRKBMN-UHFFFAOYSA-N
Synonym 4-methylphenyl chlorothioformate, o-p-tolyl chlorothioformate, p-tolyl chlorothionoformate, 4-tolyl chlorothionoformate, 4-methylphenyl chloromethanethioate, o-4-methylphenyl chlorothioformate, carbonochloridothioic acid, o-4-methylphenyl ester, o-4-methylphenyl carbonochloridothioate, chlorothioformic acid p-tolyl ester, 4-methylphenoxy methanethioyl chloride
PubChem CID 70305
IUPAC Name O-(4-methylphenyl) chloromethanethioate
SMILES CC1=CC=C(OC(Cl)=S)C=C1

Specifications

Density 1.21
Boiling Point 70°C to 72°C (1 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.574
Quantity 50 g
UN Number UN3265
Beilstein 1448986
Sensitivity Moisture sensitive
Solubility Information Miscible with common organic solvents.
Formula Weight 186.66
Percent Purity 97%
Chemical Name or Material p-Tolyl chlorothionoformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.