missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl chloroformate, 97%

Product Code. 10442651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10442651 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10442651 Supplier Thermo Scientific Acros Supplier No. 314560050
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 937-62-2 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 937-62-2
Color Colorless
Density 1.1800g/mL
Boiling Point 99.5°C (20.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H7ClO2
Linear Formula ClCO2C6H4CH3
Refractive Index 1.5080 to 1.5100
MDL Number MFCD00013255
Quantity 5 g
Specific Gravity 1.18
Synonym p-tolyl chloroformate, 4-methylphenyl chloroformate, p-tolylchloroformate, tolyl chloroformate, 4-tolylchloroformate, p-tolyl-chloroformate, 4-tolyl chloroformate, p-tolyl carbonochloridate, p-methylphenyl chloroformate
InChI Key XOFZPIYYMJUNRG-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)OC(=O)Cl
IUPAC Name (4-methylphenyl) carbonochloridate
Molecular Weight (g/mol) 170.6
PubChem CID 70304
Formula Weight 170.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material p-Tolyl chloroformate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.