missing translation for 'onlineSavingsMsg'
Learn More

p-Toluoylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog Number 11454373
16.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7391-28-8
C10H9NO
159.19
MFCD00067895
AIECDYDQPCANJK-UHFFFAOYSA-N
4-methylbenzoylacetonitrile, 4-toluoylacetonitrile, 3-4-methylphenyl-3-oxopropanenitrile, p-toluoylacetonitrile, 3-oxo-3-p-tolyl propanenitrile, 3-oxo-3-p-tolylpropanenitrile, acmc-20amdj, pubchem12072, maybridge1_004714, 4-methyl-benzoylacetonitrile
522513
3-(4-methylphenyl)-3-oxopropanenitrile
CC1=CC=C(C=C1)C(=O)CC#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7391-28-8
C10H9NO
159.19
MFCD00067895
AIECDYDQPCANJK-UHFFFAOYSA-N
4-methylbenzoylacetonitrile, 4-toluoylacetonitrile, 3-4-methylphenyl-3-oxopropanenitrile, p-toluoylacetonitrile, 3-oxo-3-p-tolyl propanenitrile, 3-oxo-3-p-tolylpropanenitrile, acmc-20amdj, pubchem12072, maybridge1_004714, 4-methyl-benzoylacetonitrile
522513
3-(4-methylphenyl)-3-oxopropanenitrile
CC1=CC=C(C=C1)C(=O)CC#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 99°C to 104°C
Quantity 1 g
Beilstein 742923
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material p-Toluoylacetonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'