missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ p-Toluoylacetonitrile, 97%

Brand:  Alfa Aesar™ A10950.14

Code : NEW

Additional Details : CAS Number : 7391-28-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11474373

  • £239.30 / 25g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

p-Toluoylacetonitrile
C10H9NO
MFCD00067895
4-Methylbenzoylacetonitrile
7391-28-8
159.19
742923

Specifications

p-Toluoylacetonitrile
25g
97%
159.19
742923
99° to 104°C
7391-28-8
C10H9NO
MFCD00067895
4-Methylbenzoylacetonitrile
Videos
SDS