missing translation for 'onlineSavingsMsg'
Learn More

p-Toluoyl Chloride, TRC

Product Code. 30453177
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30453177 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30453177

Brand: Toronto Research Chemicals TRCT536400100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Toluoyl Chloride
CAS 874-60-2
Molecular Formula C8H7ClO
Quantity 100 g
Synonym 4-Methylbenzoyl Chloride, 4-Methylbenzenecarbonyl Chloride, 4-Methylbenzoic Acid Chloride, 4-Methylbenzoyl Chloride, 4-Toluoyl Chloride, p-Methylbenzoyl Chloride, p-Toluic Acid Chloride, p-Toluyl Chloride
InChI Formula InChI=1S/C8H7ClO/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3
SMILES Cc1ccc(cc1)C(=O)Cl
IUPAC Name 4-methylbenzoyl chloride
Molecular Weight (g/mol) 154.59
Formula Weight 154.0185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.