missing translation for 'onlineSavingsMsg'
Learn More

p-Toluoyl chloride, 99%

Product Code. 11433067
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11433067

Brand: Thermo Scientific Alfa Aesar A15581.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 874-60-2 | C8H7ClO | 154.593 g/mol

p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

Solubility
Reacts with water alcohol.

Notes
Store at room temperature. Incompatible with oxidizing agent, acids, bases, active metals, alcohols, amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-60-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.593
MDL Number MFCD00000696
InChI Key NQUVCRCCRXRJCK-UHFFFAOYSA-N
Synonym p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
PubChem CID 13405
IUPAC Name 4-methylbenzoyl chloride
SMILES CC1=CC=C(C=C1)C(=O)Cl

Specifications

Melting Point -2°C
Density 1.169
Boiling Point 225°C to 227°C
Flash Point 82°C (179°F)
Assay Percent Range 99%
Odor Pungent
Refractive Index 1.553
Quantity 500 g
UN Number UN3265
Beilstein 471492
Sensitivity Moisture sensitive
Solubility Information Reacts with water alcohol.
Formula Weight 154.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material p-Toluoyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.