missing translation for 'onlineSavingsMsg'
Learn More

p-Toluoyl chloride, 98%

Product Code. 10108103 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10108103 1 L Glass bottle 1 L
10521201 5 mL Glass bottle 5mL
10666152 250 mL Glass bottle 250mL
13408378 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10108103 Supplier Thermo Scientific Acros Supplier No. 147980010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 874-60-2 | C8H7ClO | 154.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-60-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.6
MDL Number MFCD00000696
InChI Key NQUVCRCCRXRJCK-UHFFFAOYSA-N
Synonym p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
PubChem CID 13405
IUPAC Name 4-methylbenzoyl chloride
SMILES CC1=CC=C(C=C1)C(=O)Cl

Specifications

Melting Point -2.0°C
Color Colorless to Brown
Density 1.1600g/mL
Boiling Point 225.0°C to 227.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3C6H4COCl
Refractive Index 1.5525 to 1.5545
Quantity 1 L
Beilstein 09,486
Specific Gravity 1.16
Solubility Information Solubility in water: decomposes.
Formula Weight 154.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material p-Toluoyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • p-Toluoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.