missing translation for 'onlineSavingsMsg'
Learn More

p-Tolunitrile, 98+%

Product Code. 11446216
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446216 50 g 50g
11456216 250 g 250g
11436216 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446216 Supplier Thermo Scientific Alfa Aesar Supplier No. B23029.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-85-8 | C8H7N | 117.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-85-8
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.151
MDL Number MFCD00001827
InChI Key VCZNNAKNUVJVGX-UHFFFAOYSA-N
Synonym p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile
PubChem CID 7724
IUPAC Name 4-methylbenzonitrile
SMILES CC1=CC=C(C=C1)C#N

Specifications

Melting Point 26°C to 29°C
Density 0.981
Boiling Point 217°C
Flash Point 85°C (185°F)
Odor Almond-like
Quantity 50 g
Beilstein 507386
Merck Index 14,9538
Formula Weight 117.15
Percent Purity ≥98%
Chemical Name or Material p-Tolunitrile
Show More Show Less
compliance-icons
Product Identifier
  • p-Tolunitrile
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: p-Tolunitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.