missing translation for 'onlineSavingsMsg'
Learn More

p-Toluic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7081503
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
99-94-5
C8H8O2
136.15
MFCD00002565
LPNBBFKOUUSUDB-UHFFFAOYSA-N
p-toluic acid, 4-toluic acid, crithminic acid, p-methylbenzoic acid, p-toluylic acid, benzoic acid, 4-methyl, p-carboxytoluene, para-toluic acid, 4-methyl-benzoic acid, toluenecarboxylic acid
7470
CHEBI:36635
4-methylbenzoic acid
CC1=CC=C(C=C1)C(O)=O
This item is not returnable. View return policy
99-94-5
C8H8O2
136.15
MFCD00002565
LPNBBFKOUUSUDB-UHFFFAOYSA-N
p-toluic acid, 4-toluic acid, crithminic acid, p-methylbenzoic acid, p-toluylic acid, benzoic acid, 4-methyl, p-carboxytoluene, para-toluic acid, 4-methyl-benzoic acid, toluenecarboxylic acid
7470
CHEBI:36635
4-methylbenzoic acid
CC1=CC=C(C=C1)C(O)=O

It is an intermediate in some industrial processes for the oxidation of p-xylene to terephthalic acid which is used in the manufacture of polyethylene terephthalate.  It is also used in medicine, agricultural chemicals, API, dye intermediate. p-Toluic Acid is used as an intermediate for polymer stabilizers, light sensitive compounds, animal feed supplements and other organic chemicals (pharmaceuticals, pigments and dyestuff). They are also used as intermediates for the manufacture of a variety of target compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in some industrial processes for the oxidation of p-xylene to terephthalic acid which is used in the manufacture of polyethylene terephthalate.  It is also used in medicine, agricultural chemicals, API, dye intermediate. p-Toluic Acid is used as an intermediate for polymer stabilizers, light sensitive compounds, animal feed supplements and other organic chemicals (pharmaceuticals, pigments and dyestuff). They are also used as intermediates for the manufacture of a variety of target compounds.

Solubility
It is insoluble in water, but soluble in acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents, Strong bases. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Melting Point 178°C to 182°C
Density 1.06
Boiling Point 274°C
Flash Point 181°C (357°F)
Quantity 250 g
Beilstein 507600
Merck Index 14,9535
Solubility Information It is insoluble in water,but soluble in acetone.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material p-Toluic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'