missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenethiol, 98%

Product Code. 10043853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Pail
Unit Size:
100g
10g
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10043853 2.5 kg Plastic Pail 2.5kg
10113620 10 g Glass bottle 10g
10123620 100 g Glass bottle 100g
10307170 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10043853 Supplier Thermo Scientific Acros Supplier No. 138710025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-45-6 | C7H8S | 124.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-45-6
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.2
MDL Number MFCD00004851
InChI Key WLHCBQAPPJAULW-UHFFFAOYSA-N
Synonym p-toluenethiol, p-thiocresol, 4-thiocresol, 4-methylthiophenol, p-methylthiophenol, benzenethiol, 4-methyl, 4-toluenethiol, p-tolyl mercaptan, p-thiolcresol, p-tolylthiol
PubChem CID 7811
IUPAC Name 4-methylbenzenethiol
SMILES CC1=CC=C(C=C1)S

Specifications

Melting Point 40.0°C to 44.0°C
Color White to Yellow
Boiling Point 195.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic Pail
Linear Formula CH3C6H4SH
Quantity 2.5 kg
Beilstein 06, 416
Merck Index 15, 9478
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 124.2
Percent Purity 98%
Physical Form Fused Solid
Chemical Name or Material p-Toluenethiol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • p-Toluenethiol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.