missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonylacetic acid, 98%

Product Code. p-7043690
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11391507

Brand: Thermo Scientific Alfa Aesar L02102.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3937-96-0 | C9H10O4S | 214.235 g/mol

p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3937-96-0
Molecular Formula C9H10O4S
Molecular Weight (g/mol) 214.235
MDL Number MFCD00021764
InChI Key AQDHXMBUTDLAMD-UHFFFAOYSA-N
Synonym 2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o
PubChem CID 77538
IUPAC Name 2-(4-methylphenyl)sulfonylacetic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O

Specifications

Melting Point 117°C to 119°C
Quantity 5 g
Beilstein 2331311
Solubility Information Insoluble in water.
Formula Weight 214.24
Percent Purity 98%
Chemical Name or Material p-Toluenesulfonylacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.